siRNA Details
| Gene Symbol: Gm16381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166062.2 | NP_001159534.1 | 2 | 401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16381| Gene Symbol: Gm16381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166062.2 | NP_001159534.1 | 2 | 401 | ||