siRNA Details
| Gene Symbol: Gm6812 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098842.2 | NP_001092312.1 | 2 | 309 | ||
| Gene Symbol: 1700020N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029316.1 | NP_083592.1 | 2 | 328 | ||
| GenBank mRNA | AK006180.1 | - | 2 | 310 | ||
| BC049765.1 | - | 306 | ||||
| CA464409.1 | - | 2 | 330 | |||

Gm6812
Genomic Map