siRNA Details
| Gene Symbol: RPS4Y1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008.3 | NP_000999.1 | 7 | 785 | ||
| XM_017030061.1 | XP_016885550.1 | 7 | 869 | |||
| GenBank mRNA | AK222594.1 | - | 7 | 766 | ||
| AK292819.1 | - | 7 | 770 | |||
| BC010286.1 | - | 6 | 754 | 3048929 | ||
| BC100905.2 | - | 6 | 742 | 40003602 | ||
| BC100906.1 | - | 6 | 740 | |||
| M58459.1 | - | 6 | 754 | |||

RPS4Y1
Genomic Map