siRNA Details
| Gene Symbol: Fbxo39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099688.2 | NP_001093158.1 | 2 | 411 | ||
| GenBank mRNA | BU937284.1 | - | 2 | 414 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo39| Gene Symbol: Fbxo39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099688.2 | NP_001093158.1 | 2 | 411 | ||
| GenBank mRNA | BU937284.1 | - | 2 | 414 | ||