siRNA Details
| Gene Symbol: Gm14410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078446.1 | - | 770 | |||
| Gene Symbol: Gm14322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243903.1 | NP_001230832.1 | 5 | 1090 | ||

Gm14410
Genomic Map