siRNA Details
| Gene Symbol: Gm38425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251890.1 | NP_001238819.1 | 2 | 531 | ||
| GenBank mRNA | DV654665.1 | - | 2 | 531 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm38425| Gene Symbol: Gm38425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001251890.1 | NP_001238819.1 | 2 | 531 | ||
| GenBank mRNA | DV654665.1 | - | 2 | 531 | ||