siRNA Details
| Gene Symbol: Phf11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164289.1 | NP_001157761.1 | 7 | 998 | ||
| GenBank mRNA | EU155106.1 | - | 7 | 2868 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf11c| Gene Symbol: Phf11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164289.1 | NP_001157761.1 | 7 | 998 | ||
| GenBank mRNA | EU155106.1 | - | 7 | 2868 | ||