siRNA Details
Gene Symbol: Phf11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164289.1 | NP_001157761.1 | 2, 3 | 598 | ||
GenBank mRNA | EU155106.1 | - | 2, 3 | 2468 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Phf11c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164289.1 | NP_001157761.1 | 2, 3 | 598 | ||
GenBank mRNA | EU155106.1 | - | 2, 3 | 2468 |