siRNA Details
Gene Symbol: Cldn34-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101585.1 | NP_001095055.1 | 1 | 532 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cldn34-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101585.1 | NP_001095055.1 | 1 | 532 |