siRNA Details
| Gene Symbol: Vmn2r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104648.1 | NP_001098118.1 | 2 | 604 | ||
| GenBank mRNA | AB919109.1 | - | 2 | 604 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r56| Gene Symbol: Vmn2r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104648.1 | NP_001098118.1 | 2 | 604 | ||
| GenBank mRNA | AB919109.1 | - | 2 | 604 | ||
Set Membership: |
|