siRNA Details
Gene Symbol: Heatr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045543.2 | NP_001039008.1 | 5 | 539 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Heatr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045543.2 | NP_001039008.1 | 5 | 539 |