siRNA Details
| Gene Symbol: Spint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177401.1 | NP_001170872.1 | 2 | 251 | ||
| GenBank mRNA | AK136730.1 | - | 2 | 229 | ||
| BU938823.1 | - | 2 | 254 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spint3| Gene Symbol: Spint3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177401.1 | NP_001170872.1 | 2 | 251 | ||
| GenBank mRNA | AK136730.1 | - | 2 | 229 | ||
| BU938823.1 | - | 2 | 254 | |||