siRNA Details
| Gene Symbol: Gm7008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB273562.1 | - | 3 | 303 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm7008| Gene Symbol: Gm7008 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | CB273562.1 | - | 3 | 303 | ||