siRNA Details
| Gene Symbol: Krtap1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039502.2 | NP_001034591.1 | 1 | 883 | ||
| GenBank mRNA | AK131904.1 | - | 1 | 884 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap1-4| Gene Symbol: Krtap1-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039502.2 | NP_001034591.1 | 1 | 883 | ||
| GenBank mRNA | AK131904.1 | - | 1 | 884 | ||