siRNA Details
| Gene Symbol: TTK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166691.1 | NP_001160163.1 | 15 | 1819 | ||
| NM_003318.4 | NP_003309.2 | 15 | 1810 | |||
| XM_011536099.2 | XP_011534401.1 | 15 | 1784 | |||
| XM_011536100.2 | XP_011534402.1 | 15 | 1807 | |||
| XM_017011242.1 | XP_016866731.1 | 16 | 2339 | |||
| GenBank mRNA | AK095599.1 | - | 15 | 2279 | ||
| AK292460.1 | - | 15 | 1782 | |||
| AK315696.1 | - | 15 | 1727 | |||
| AX775733.1 | - | 15 | 1782 | |||
| BC000633.2 | - | 15 | 1773 | 3343925 | ||
| BC032858.2 | - | 15 | 1775 | 5270012 | ||
| X70500.1 | - | 2 | 232 | |||

TTK
Genomic Map