siRNA Details
| Gene Symbol: RPS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017.2 | NP_001008.1 | 5 | 416 | ||
| GenBank mRNA | AK312060.1 | - | 5 | 410 | ||
| BC000475.1 | - | 1 | 395 | 2961637 | ||
| BC006772.1 | - | 1 | 397 | 2899987 | ||
| BC029732.1 | - | 5 | 396 | 4388273 | ||
| BC066322.1 | - | 5 | 410 | 4816284 | ||
| BC100032.1 | - | 5 | 396 | 6025884 | ||
| L01124.1 | - | 5 | 416 | |||
| X79239.1 | - | 5 | 384 | |||

RPS13
Genomic Map