siRNA Details
| Gene Symbol: Igh-VJ558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC003844.1 | - | 2 | 181 | 3497357 | |
| Gene Symbol: Ighv9-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF045485.1 | - | 135 | |||
| AF045486.1 | - | 135 | ||||

Igh-VJ558
Genomic Map