siRNA Details
| Gene Symbol: Eif4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007917.3 | NP_031943.3 | 4 | 317 | ||
| GenBank mRNA | AK082867.1 | - | 3 | 198 | ||
| AK146757.1 | - | 3 | 197 | |||
| AK167097.1 | - | 3 | 206 | |||
| BC010759.1 | - | 3 | 191 | 4216609 | ||
| BC085087.1 | - | 3 | 189 | 6438419 | ||
| CN714483.1 | - | 4 | 317 | |||
| M61731.1 | - | 3 | 203 | |||

Eif4e
Genomic Map