siRNA Details
| Gene Symbol: Zfp808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 1248 | ||
| GenBank mRNA | BC061203.1 | - | 4 | 1248 | 30248220 | |
| Gene Symbol: Zfp934 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162911.1 | NP_001156383.1 | 4 | 1222 | ||
| GenBank mRNA | BC021319.1 | - | 1141 | |||
| BC115861.1 | - | 3 | 1073 | |||

Zfp808
Genomic Map