siRNA Details
| Gene Symbol: Trim43c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177858.1 | NP_001171329.1 | 7 | 1478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim43c| Gene Symbol: Trim43c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177858.1 | NP_001171329.1 | 7 | 1478 | ||