siRNA Details
| Gene Symbol: Nid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010917.2 | NP_035047.2 | 18 | 3560 | ||
| GenBank mRNA | AK084876.1 | - | 18 | 3499 | ||
| AK144878.1 | - | 18 | 3525 | |||
| AK166779.1 | - | 18 | 3496 | |||
| BC052939.1 | - | 18 | 3555 | |||
| BC131668.1 | - | 5 | 736 | |||
| BC131669.1 | - | 18 | 3471 | 40110859 | ||
| X14194.1 | - | 18 | 3471 | |||
| X14480.1 | - | 18 | 3481 | |||

Nid1
Genomic Map