siRNA Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199210.1 | NP_001186139.1 | 8 | 1990 | ||
| NM_027627.2 | NP_081903.1 | 7 | 1846 | |||
| GenBank mRNA | AK037486.1 | - | 6 | 1701 | ||
| BC132305.1 | - | 8 | 1990 | 40130582 | ||
| BC144751.1 | - | 7 | 1792 | |||
| BC157969.1 | - | 6 | 1540 | 9007291 | ||

Eva1c
Genomic Map