siRNA Details
| Gene Symbol: Eva1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199210.1 | NP_001186139.1 | 8 | 1958 | ||
| NM_027627.2 | NP_081903.1 | 7 | 1814 | |||
| GenBank mRNA | BC132305.1 | - | 8 | 1958 | 40130582 | |
| BC144751.1 | - | 7 | 1760 | |||
| BC157969.1 | - | 6 | 1508 | 9007291 | ||

Eva1c
Genomic Map