siRNA Details
Gene Symbol: RPS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022551.2 | NP_072045.1 | 2 | 113 | ||
GenBank mRNA | AY536376.1 | - | 2 | 92 | ||
BC101786.1 | - | 2 | 108 | 8069292 | ||
BC101788.1 | - | 2 | 108 | 8069294 | ||
BC106063.1 | - | 2 | 166 | 4386992 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011296, BC091729, AK159437, M76763, AK146354, and others.
Panther Classification:
Gene Ontology Categories:
