siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132279.1 | - | 694 | |||
| BC026442.1 | - | 678 | ||||
| BC037657.1 | - | 653 | ||||
| BC046825.1 | - | 690 | ||||
| BC094629.1 | - | 7 | 714 | 3376726 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb8os| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132279.1 | - | 694 | |||
| BC026442.1 | - | 678 | ||||
| BC037657.1 | - | 653 | ||||
| BC046825.1 | - | 690 | ||||
| BC094629.1 | - | 7 | 714 | 3376726 | ||