siRNA Details
Gene Symbol: Gm12250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 385 | ||
GenBank mRNA | DQ508486.1 | - | 1 | 123 | ||
DQ508487.1 | - | 1 | 123 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm12250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135115.1 | NP_001128587.1 | 2 | 385 | ||
GenBank mRNA | DQ508486.1 | - | 1 | 123 | ||
DQ508487.1 | - | 1 | 123 |