siRNA Details
Gene Symbol: Lonrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081150.1 | NP_001074619.1 | 12 | 3489 | ||
GenBank mRNA | BC094431.1 | - | 9 | 2295 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lonrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081150.1 | NP_001074619.1 | 12 | 3489 | ||
GenBank mRNA | BC094431.1 | - | 9 | 2295 |