siRNA Details
Gene Symbol: Vmn1r170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166722.1 | NP_001160194.1 | 1 | 150 |
Gene Symbol: Vmn1r167 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101562.1 | NP_001095032.1 | 1 | 183 |
Gene Symbol: Vmn1r169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166843.1 | NP_001160315.1 | 1 | 150 |
Gene Symbol: Vmn1r171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030737.2 | NP_109662.1 | 1 | 550 | ||
GenBank mRNA | AF324871.1 | - | 560 | |||
BC111911.1 | - | 1 | 550 | 40046770 |
Gene Symbol: Vmn1r175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166727.1 | NP_001160199.1 | 1 | 150 |
Gene Symbol: Vmn1r176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166721.1 | NP_001160193.1 | 1 | 150 |
