siRNA Details
| Gene Symbol: Vmn1r169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166843.1 | NP_001160315.1 | 1 | 864 | ||
| Gene Symbol: Vmn1r176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166721.1 | NP_001160193.1 | 1 | 864 | ||

Vmn1r169
Genomic Map