siRNA Details
| Gene Symbol: Arl4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177305.4 | NP_796279.2 | 1 | 609 | ||
| GenBank mRNA | AK080335.1 | - | 1 | 553 | ||
| AK080380.1 | - | 1 | 553 | |||
| AK080743.1 | - | 1 | 562 | |||
| AK169890.1 | - | 1 | 46 | |||
| BC049804.1 | - | 294 | ||||
| BC055769.1 | - | 1 | 569 | 6414771 | ||

Arl4c
Genomic Map