siRNA Details
| Gene Symbol: RPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030001.2 | NP_001025172.1 | 1, 2 | 135 | ||
| NM_001032.4 | NP_001023.1 | 1, 2 | 135 | |||
| GenBank mRNA | AK026649.1 | - | 1 | 95 | ||
| AK307833.1 | - | 1, 2 | 96 | |||
| BC015974.1 | - | 1, 2 | 99 | 4105424 | ||
| BC032813.1 | - | 1 | 70 | 5217083 | ||
| BC035313.1 | - | 70 | 3868721 | |||
| BF128896.1 | - | 1, 2 | 42 | |||
| BQ433530.1 | - | 1, 2 | 79 | |||
| BU942244.1 | - | 1, 2 | 138 | |||
| L31610.1 | - | 1, 2 | 76 | |||
| U14973.1 | - | 1, 2 | 95 | |||

RPS29
Genomic Map