siRNA Details
| Gene Symbol: Tma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183250.2 | NP_899073.1 | 4 | 613 | ||
| GenBank mRNA | AK003762.1 | - | 4 | 614 | ||
| AK169860.1 | - | 4 | 607 | |||
| BC054363.1 | - | 4 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tma7| Gene Symbol: Tma7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183250.2 | NP_899073.1 | 4 | 613 | ||
| GenBank mRNA | AK003762.1 | - | 4 | 614 | ||
| AK169860.1 | - | 4 | 607 | |||
| BC054363.1 | - | 4 | 585 | |||