siRNA Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177185.4 | NP_796159.3 | 11 | 2113 | ||
| GenBank mRNA | AK051609.1 | - | 11 | 1486 | ||
| BC043095.1 | - | 1358 | ||||
| BC044744.1 | - | 6 | 815 | |||
| BC051458.1 | - | 11 | 2088 | 6468017 | ||

Ubn2
Genomic Map