siRNA Details
| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 6, 7 | 491 | ||
| GenBank mRNA | AK003561.1 | - | 5, 6 | 450 | ||
| BC033045.1 | - | 5, 6 | 469 | 1068150 | ||
| BC094619.1 | - | 6, 7 | 478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcf1| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 6, 7 | 491 | ||
| GenBank mRNA | AK003561.1 | - | 5, 6 | 450 | ||
| BC033045.1 | - | 5, 6 | 469 | 1068150 | ||
| BC094619.1 | - | 6, 7 | 478 | |||