siRNA Details
| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 3 | 120 | ||
| GenBank mRNA | AK003561.1 | - | 2 | 79 | ||
| BC033045.1 | - | 2 | 98 | 1068150 | ||
| BC094619.1 | - | 3 | 107 | |||
| CJ085020.1 | - | 3 | 135 | |||

Fcf1
Genomic Map