siRNA Details
| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 7 | 517 | ||
| GenBank mRNA | AK003561.1 | - | 6 | 476 | ||
| BC094619.1 | - | 7 | 504 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcf1| Gene Symbol: Fcf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028632.2 | NP_082908.2 | 7 | 517 | ||
| GenBank mRNA | AK003561.1 | - | 6 | 476 | ||
| BC094619.1 | - | 7 | 504 | |||