siRNA Details
| Gene Symbol: Gm6225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086784.1 | - | 2 | 1168 | ||
| AK163460.1 | - | 2 | 1151 | |||
| Gene Symbol: Gm1971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK143472.1 | - | 1 | 768 | ||
| AK165166.1 | - | 1 | 768 | |||
| AK166762.1 | - | 1 | 768 | |||

Gm6225
Genomic Map