siRNA Details
Gene Symbol: Gm6225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086784.1 | - | 2 | 850 | ||
AK163460.1 | - | 2 | 833 | |||
AK166419.1 | - | 2 | 839 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm6225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK086784.1 | - | 2 | 850 | ||
AK163460.1 | - | 2 | 833 | |||
AK166419.1 | - | 2 | 839 |