siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 8 | 1761 | ||
| NM_027280.3 | NP_081556.3 | 10 | 1971 | |||
| GenBank mRNA | AK020249.1 | - | 1 | 50 | ||
| AK076164.1 | - | 10 | 1963 | |||
| AK082367.1 | - | 10 | 1960 | |||
| AK193660.1 | - | 1 | 219 | |||

Nkd1
Genomic Map