siRNA Details
| Gene Symbol: Col6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167923.1 | NP_001161395.1 | 4 | 1377 | ||
| GenBank mRNA | AM748256.1 | - | 4 | 1377 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col6a5| Gene Symbol: Col6a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167923.1 | NP_001161395.1 | 4 | 1377 | ||
| GenBank mRNA | AM748256.1 | - | 4 | 1377 | ||