siRNA Details
| Gene Symbol: Mettl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176917.4 | NP_795891.2 | 2 | 845 | ||
| GenBank mRNA | AK043389.1 | - | 2 | 1124 | ||
| AK080683.1 | - | 2 | 1195 | |||
| AK157268.1 | - | 2 | 846 | |||
| BC099943.1 | - | 2 | 1325 | 30020588 | ||
| BC115977.1 | - | 1 | 648 | 40105192 | ||

Mettl4
Genomic Map