siRNA Details
| Gene Symbol: Nup37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027191.2 | NP_081467.2 | 6, 7 | 676 | ||
| NM_028334.4 | NP_082610.1 | 6, 7 | 694 | |||
| GenBank mRNA | AK010370.1 | - | 6, 7 | 620 | ||
| AK012890.1 | - | 6, 7 | 690 | |||
| AK040463.1 | - | 6, 7 | 677 | |||
| AK165537.1 | - | 6, 7 | 640 | |||
| BC011102.1 | - | 6, 7 | 660 | 3982076 | ||
| BC032180.1 | - | 5, 6 | 604 | 5151847 | ||

Nup37
Genomic Map