siRNA Details
| Gene Symbol: Fam81b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AU023564.1 | - | 1 | 121 | ||
| BB615662.1 | - | 2 | 106 | |||
| BG068689.2 | - | 1 | 126 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam81b| Gene Symbol: Fam81b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AU023564.1 | - | 1 | 121 | ||
| BB615662.1 | - | 2 | 106 | |||
| BG068689.2 | - | 1 | 126 | |||