siRNA Details
| Gene Symbol: Gbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039647.2 | NP_001034736.2 | 6 | 884 | ||
| GenBank mRNA | BC111039.1 | - | 6 | 881 | 5349539 | |
| EU304258.1 | - | 5 | 801 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gbp11| Gene Symbol: Gbp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039647.2 | NP_001034736.2 | 6 | 884 | ||
| GenBank mRNA | BC111039.1 | - | 6 | 881 | 5349539 | |
| EU304258.1 | - | 5 | 801 | |||