siRNA Details
| Gene Symbol: Ube2n | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080560.3 | NP_542127.1 | 3 | 430 | ||
| GenBank mRNA | AK005302.1 | - | 3 | 448 | ||
| AK005788.1 | - | 3 | 440 | |||
| AK161968.1 | - | 3 | 446 | |||
| AY039837.1 | - | 3 | 439 | |||
| BC034898.1 | - | 3 | 407 | 1382079 | ||
| BC052203.1 | - | 3 | 334 | |||
| BC067069.1 | - | 3 | 430 | 30550024 | ||
| BC132630.1 | - | 3 | 405 | 40130907 | ||
| BC138211.1 | - | 3 | 405 | 8861230 | ||
| Y09873.1 | - | 3 | 417 | |||

Ube2n
Genomic Map