siRNA Details
| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137547.1 | NP_001131019.1 | 2 | 1484 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp51| Gene Symbol: Usp51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001137547.1 | NP_001131019.1 | 2 | 1484 | ||