siRNA Details
| Gene Symbol: Cldn34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199311.1 | NP_001186240.1 | 1 | 364 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cldn34a| Gene Symbol: Cldn34a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199311.1 | NP_001186240.1 | 1 | 364 | ||