siRNA Details
| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013901.2 | NP_038929.2 | 4 | 965 | ||
| GenBank mRNA | AK146175.1 | - | 4 | 957 | ||
| AK159290.1 | - | 4 | 822 | |||
| BC005474.1 | - | 4 | 927 | 3591856 | ||
| BC132461.1 | - | 3 | 902 | 40130738 | ||
| BC138459.1 | - | 3 | 902 | 8861479 | ||
| BC147738.1 | - | 3 | 903 | 9056687 | ||
| BC147744.1 | - | 3 | 903 | 9056693 | ||

Slc39a1
Genomic Map