siRNA Details
| Gene Symbol: Slc39a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013901.2 | NP_038929.2 | 4 | 1298 | ||
| GenBank mRNA | AK146175.1 | - | 4 | 1290 | ||
| AK159290.1 | - | 4 | 1155 | |||
| BC132461.1 | - | 3 | 1235 | 40130738 | ||
| BC138459.1 | - | 3 | 1235 | 8861479 | ||

Slc39a1
Genomic Map