siRNA Details
| Gene Symbol: Gm7168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122977.1 | NP_001116449.1 | 1 | 426 | ||
| GenBank mRNA | BC107382.2 | - | 1 | 426 | 40057241 | |
| BC125014.1 | - | 1 | 426 | 40057242 | ||
| BC125267.1 | - | 1 | 426 | 40086171 | ||

Gm7168
Genomic Map